2,6-Diazaspiro[3.4]octan-5-one hydrochloride

Lieferant: Apollo Scientific

OR960531-1G
APOSOR960531-1GEA 1710 EUR
APOSOR960531-1G
2,6-Diazaspiro[3.4]octan-5-one hydrochloride
2,6-Diazaspiro[3.4]octan-5-one hydrochloride
Formel: C₆H₁₀N₂O·ClH
Molecular Weight: 162.617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053741
CAS-Nummer: 1203683-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR