Boc-D-Chg-OH 97%
Lieferant: Apollo Scientific
Synonyme:
Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine
OR960787-1G
OR960787-25G
OR960787-5G
APOSOR960787-1GEA
52.2
EUR
APOSOR960787-1G
APOSOR960787-25G
APOSOR960787-5G
Boc-D-Chg-OH 97%
Boc-D-Chg-OH
Formel:
C₁₃H₂₃NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00133629 CAS-Nummer: 70491-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...