3,5-Dibrom-1H-1,2,4-triazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dibromo-1,2,4-triazole
OR9608-25G
OR9608-5G
APOSOR9608-25GEA
46.4
EUR
APOSOR9608-25G
APOSOR9608-5G
3,5-Dibrom-1H-1,2,4-triazol 98%
3,5-Dibrom-1H-1,2,4-triazol
Formel:
C₂HBr₂N₃ Molecular Weight: 226.8600 g/mol Schmelzpunkt: 201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00487260 CAS-Nummer: 7411-23-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...