N-Fmoc-N-methyl-D-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-N-Me-Phe-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid

OR960818-1G OR960818-500MG OR960818-5G
APOSOR960818-1GEA 126 EUR
APOSOR960818-1G APOSOR960818-500MG APOSOR960818-5G
N-Fmoc-N-methyl-D-phenylalanine 97%
N-Fmoc-N-methyl-D-phenylalanine
Formel: C₂₅H₂₃NO₄
Molecular Weight: 401.462 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153392
CAS-Nummer: 138775-05-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR