6-Ethoxyimidazo[1,2-b]pyridazine 95%

Lieferant: Apollo Scientific

OR960828-1G OR960828-250MG
APOSOR960828-1GEA 364 EUR
APOSOR960828-1G APOSOR960828-250MG
6-Ethoxyimidazo[1,2-b]pyridazine 95%
6-Ethoxyimidazo[1,2-b]pyridazine
Formel: C₈H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18782899
CAS-Nummer: 57470-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR