Chloromethyl benzoate 98%

Lieferant: Apollo Scientific

OR960834-25G
APOSOR960834-25GEA 296 EUR
APOSOR960834-25G
Chloromethyl benzoate 98%
Chloromethyl benzoate
Formel: C₈H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00040116
CAS-Nummer: 5335-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR