BOC-N-Methyl-D-Alanin 97%

Lieferant: Apollo Scientific

Synonyme: BOC-N-methyl-D-alanine, (R)-2-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid, Boc-N-Me-D-Ala-OH, N-Boc-N-methyl-D-alanine

OR960889-25G
APOSOR960889-25GEA 65.4 EUR
APOSOR960889-25G
BOC-N-Methyl-D-Alanin 97%
BOC-N-Methyl-D-Alanin
Formel: C₉H₁₇NO₄
Molecular Weight: 203.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063137
CAS-Nummer: 19914-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR