Isoindoline-5-carboxylic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR960892-250MG
APOSOR960892-250MGEA 112 EUR
APOSOR960892-250MG
Isoindoline-5-carboxylic acid hydrochloride 95%
Isoindoline-5-carboxylic acid hydrochloride
Formel: C₉H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10699395
CAS-Nummer: 149353-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR