2-(4-Methylthiazol-5-yl)ethylamine 97%

Lieferant: Apollo Scientific

OR960927-1G OR960927-5G
APOSOR960927-1GEA 203 EUR
APOSOR960927-1G APOSOR960927-5G
2-(4-Methylthiazol-5-yl)ethylamine 97%
Formel: C₆H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08060722
CAS-Nummer: 58981-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR