Methyl-7-azaindole-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate

OR960932-10G OR960932-1G OR960932-5G
APOSOR960932-10GEA 230 EUR
APOSOR960932-10G APOSOR960932-1G APOSOR960932-5G
Methyl-7-azaindole-2-carboxylate 98%
Methyl-7-azaindole-2-carboxylate
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08752938
CAS-Nummer: 394223-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR