Methyl-5-methylthiazole-4-carboxylate 95%
Lieferant: Apollo Scientific
OR960944-1G
OR960944-250MG
APOSOR960944-1GEA
719
EUR
APOSOR960944-1G
APOSOR960944-250MG
Methyl-5-methylthiazole-4-carboxylate 95%
Methyl-5-methylthiazole-4-carboxylate
Formel:
C₆H₇NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109386 CAS-Nummer: 68751-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...