Methyl-5-methylthiazole-4-carboxylate 95%

Lieferant: Apollo Scientific

OR960944-1G OR960944-250MG
APOSOR960944-1GEA 719 EUR
APOSOR960944-1G APOSOR960944-250MG
Methyl-5-methylthiazole-4-carboxylate 95%
Methyl-5-methylthiazole-4-carboxylate
Formel: C₆H₇NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11109386
CAS-Nummer: 68751-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR