(S)-Methyl-2-aminohexanoate hydrochloride 98%

Lieferant: Apollo Scientific

OR960947-100G OR960947-25G OR960947-5G
APOSOR960947-100GEA 0 EUR
APOSOR960947-100G APOSOR960947-25G APOSOR960947-5G
(S)-Methyl-2-aminohexanoate hydrochloride 98%
(S)-Methyl-2-aminohexanoate hydrochloride
Formel: C₇H₁₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00077151
CAS-Nummer: 3844-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR