Fmoc-3-Cyclohexyl-L-Alanin 97%

Lieferant: Apollo Scientific

OR960948-1G OR960948-25G OR960948-5G
APOSOR960948-1GEA 0 EUR
APOSOR960948-1G APOSOR960948-25G APOSOR960948-5G
Fmoc-3-Cyclohexyl-L-Alanin 97%
Fmoc-3-Cyclohexyl-L-Alanin
Formel: C₂₄H₂₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00065614
CAS-Nummer: 135673-97-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR