Fmoc-3-Cyclohexyl-L-Alanin 97%
Lieferant: Apollo Scientific
OR960948-1G
OR960948-25G
OR960948-5G
APOSOR960948-1GEA
0
EUR
APOSOR960948-1G
APOSOR960948-25G
APOSOR960948-5G
Fmoc-3-Cyclohexyl-L-Alanin 97%
Fmoc-3-Cyclohexyl-L-Alanin
Formel:
C₂₄H₂₇NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065614 CAS-Nummer: 135673-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...