N-Me-Val-OMe·HCl 95%

Lieferant: Apollo Scientific

Synonyme: N-Me-Val-OMe HCl, N-Methyl-L-valine methyl ester hydrochloride, N-Me-Val-OMe·HCl

OR961005-1G
APOSOR961005-1GEA 23.2 EUR
APOSOR961005-1G
N-Me-Val-OMe·HCl 95%
N-Me-Val-OMe·HCl
Formel: C₇H₁₆ClNO₂
Molecular Weight: 181.66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00238325
CAS-Nummer: 3339-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR