Boc-L-Asp(OAll)-OH

Lieferant: Apollo Scientific

Synonyme: Boc-Asp(OAll)-OH, (S)-4-(Allyloxy)-2-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid

OR961039-25G OR961039-5G OR961039-100G
APOSOR961039-25GEA 302 EUR
APOSOR961039-25G APOSOR961039-5G APOSOR961039-100G
Boc-L-Asp(OAll)-OH
Boc-L-Asp(OAll)-OH
Formel: C₁₂H₁₉NO₆
Schmelzpunkt: 44…50 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00273398
CAS-Nummer: 132286-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR