Boc-L-Asp(OAll)-OH
Lieferant: Apollo Scientific
Synonyme:
Boc-Asp(OAll)-OH, (S)-4-(Allyloxy)-2-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid
OR961039-25G
OR961039-5G
OR961039-100G
APOSOR961039-25GEA
302
EUR
APOSOR961039-25G
APOSOR961039-5G
APOSOR961039-100G
Boc-L-Asp(OAll)-OH
Boc-L-Asp(OAll)-OH
Formel:
C₁₂H₁₉NO₆ Schmelzpunkt: 44…50 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00273398 CAS-Nummer: 132286-77-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...