2-Naphthalen-2-yl-ethylamine 96%

Lieferant: Apollo Scientific

OR961210-1G OR961210-250MG OR961210-5G
APOSOR961210-1GEA 290 EUR
APOSOR961210-1G APOSOR961210-250MG APOSOR961210-5G
2-Naphthalen-2-yl-ethylamine 96%
2-Naphthalen-2-yl-ethylamine
Formel: C₁₂H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD01529868
CAS-Nummer: 2017-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR