3-Cyano-1H-indazole 98%

Lieferant: Apollo Scientific
Achtung

OR961248-1G
APOSOR961248-1GEA 43.5 EUR
APOSOR961248-1G
3-Cyano-1H-indazole 98%
3-Cyano-1H-indazole
Formel: C₈H₅N₃
Lagertemperatur: Kühlschrank
MDL: MFCD01860240
CAS-Nummer: 50264-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR