(6-Methyl-1H-benzimidazol-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR961343-1G
APOSOR961343-1GEA 306 EUR
APOSOR961343-1G
(6-Methyl-1H-benzimidazol-2-yl)methanol 95%
(6-Methyl-1H-benzimidazol-2-yl)methanol
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD01874328
CAS-Nummer: 20034-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR