3-(Boc-amino)-2-phenylpropanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 3-((tert-Butoxycarbonyl)amino)-2-phenylpropanoic acid

OR961404-1G
APOSOR961404-1GEA 410 EUR
APOSOR961404-1G
3-(Boc-amino)-2-phenylpropanoic acid 97%
3-(Boc-amino)-2-phenylpropanoic acid
Formel: C₁₄H₁₉NO₄
Molecular Weight: 265.305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03003692
CAS-Nummer: 67098-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR