7-Ethyl-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

OR961407-1G
APOSOR961407-1GEA 319 EUR
APOSOR961407-1G
7-Ethyl-1H-indole-3-carbaldehyde 95%
7-Ethyl-1H-indole-3-carbaldehyde
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03043379
CAS-Nummer: 154989-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR