Quinolin-5-ylmethanol 95%
Lieferant: Apollo Scientific
OR961517-1G
APOSOR961517-1GEA
464
EUR
APOSOR961517-1G
Quinolin-5-ylmethanol 95%
Quinolin-5-ylmethanol
Formel:
C₁₀H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160574 CAS-Nummer: 16178-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...