(R)-3-Amino-3-(2-bromophenyl)propionic acid 97%

Lieferant: Apollo Scientific

OR961588-1G OR961588-500MG
APOSOR961588-1GEA 348 EUR
APOSOR961588-1G APOSOR961588-500MG
(R)-3-Amino-3-(2-bromophenyl)propionic acid 97%
(R)-3-Amino-3-(2-bromophenyl)propionic acid
Formel: C₉H₁₀BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04113641
CAS-Nummer: 737751-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR