2-Chloro-4-phenylpyridine 98%

Lieferant: Apollo Scientific

OR961595-10G OR961595-1G OR961595-5G
APOSOR961595-10GEA 528 EUR
APOSOR961595-10G APOSOR961595-1G APOSOR961595-5G
2-Chloro-4-phenylpyridine 98%
2-Chloro-4-phenylpyridine
Formel: C₁₁H₈ClN
Molecular Weight: 189.6400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114163
CAS-Nummer: 42260-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR