Methyl-1-aminocyclobutanecarboxylate hydrochloride

Lieferant: Apollo Scientific

OR961598-25G OR961598-5G
APOSOR961598-25GEA 0 EUR
APOSOR961598-25G APOSOR961598-5G
Methyl-1-aminocyclobutanecarboxylate hydrochloride
Methyl-1-aminocyclobutanecarboxylate hydrochloride
Formel: C₆H₁₂ClNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD04115288
CAS-Nummer: 92398-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR