1-Benzyl-pyrrolidine 98%

Lieferant: Apollo Scientific

OR961673-1G OR961673-5G
APOSOR961673-1GEA 24.4 EUR
APOSOR961673-1G APOSOR961673-5G
1-Benzyl-pyrrolidine 98%
1-Benzyl-pyrrolidine
Formel: C₁₁H₁₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD00127807
CAS-Nummer: 29897-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR