5-Bromo-2-formylbenzonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-cyanobenzaldehyde

OR961690-250MG
APOSOR961690-250MGEA 59.6 EUR
APOSOR961690-250MG
5-Bromo-2-formylbenzonitrile 97%
5-Bromo-2-formylbenzonitrile
Formel: C₈H₄BrNO
Molecular Weight: 210,027 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261030
CAS-Nummer: 523977-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR