Sodium pivalate 95%

Lieferant: Apollo Scientific
Achtung

OR961732-25G OR961732-5G
APOSOR961732-25GEA 82.8 EUR
APOSOR961732-25G APOSOR961732-5G
Sodium pivalate 95%
Sodium pivalate
Formel: C₅H₉O₂·Na
Lagertemperatur: Raumtemperatur
MDL: MFCD00064202
CAS-Nummer: 1184-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR