(2,4-Dichloro-phenyl)-ethyl-amine 95%

Lieferant: Apollo Scientific

OR961773-1G
APOSOR961773-1GEA 609 EUR
APOSOR961773-1G
(2,4-Dichloro-phenyl)-ethyl-amine 95%
(2,4-Dichloro-phenyl)-ethyl-amine
Formel: C₈H₉Cl₂N
Molecular Weight: 190,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658422
CAS-Nummer: 35113-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR