H-Cys(StBu)-OH

Lieferant: Apollo Scientific

OR961825-5G
APOSOR961825-5GEA 0 EUR
APOSOR961825-5G
H-Cys(StBu)-OH
H-Cys(StBu)-OH
Formel: C₇H₁₅NO₂S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00002609
CAS-Nummer: 30044-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR