2-Methoxy-4,6-dimethylpyrimidine 95%

Lieferant: Apollo Scientific

OR961880-1G
APOSOR961880-1GEA 596 EUR
APOSOR961880-1G
2-Methoxy-4,6-dimethylpyrimidine 95%
2-Methoxy-4,6-dimethylpyrimidine
Formel: C₇H₁₀N₂O
Molecular Weight: 138,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02765642
CAS-Nummer: 14001-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR