Fmoc-3-cyano-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: Fmoc-Phe(3-CN)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-cyanophenyl)propanoic acid

OR961964-5G
APOSOR961964-5GEA 126 EUR
APOSOR961964-5G
Fmoc-3-cyano-L-phenylalanine 97%
Fmoc-3-cyano-L-phenylalanine
Formel: C₂₅H₂₀N₂O₄
Molecular Weight: 412.45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00797585
CAS-Nummer: 205526-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR