1-Methyl-4-nitroimidazole 95%

Lieferant: Apollo Scientific

OR961982-5G
APOSOR961982-5GEA 154 EUR
APOSOR961982-5G
1-Methyl-4-nitroimidazole 95%
1-Methyl-4-nitroimidazole
Formel: C₄H₅N₃O₂
Molecular Weight: 127.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159683
CAS-Nummer: 3034-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR