Allyl-4-oxopiperidine-1-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: 1-N-Alloc-4-piperidone

OR962024-1G
APOSOR962024-1GEA 328 EUR
APOSOR962024-1G
Allyl-4-oxopiperidine-1-carboxylate 95%
Allyl-4-oxopiperidine-1-carboxylate
Formel: C₉H₁₃NO₃
Molecular Weight: 183.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02183578
CAS-Nummer: 306296-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR