3-(5-Bromothien-2-yl)-L-alanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(5-bromothiophen-2-yl)propanoic acid, H-2-(5-Br-Thienyl)-Ala-OH

OR962048-1G
APOSOR962048-1GEA 133 EUR
APOSOR962048-1G
3-(5-Bromothien-2-yl)-L-alanine 97%
3-(5-Bromothien-2-yl)-L-alanine
Formel: C₇H₈BrNO₂S
Molecular Weight: 250.113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04117834
CAS-Nummer: 154593-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR