4-Methylphthalimide 95%

Lieferant: Apollo Scientific

Synonyme: 5-Methylisoindoline-1,3-dione

OR962271-5G
APOSOR962271-5GEA 69.6 EUR
APOSOR962271-5G
4-Methylphthalimide 95%
4-Methylphthalimide
Formel: C₉H₇NO₂
Molecular Weight: 161,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05155224
CAS-Nummer: 40314-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR