(S)-N-Fmoc-azetidine-2-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid, Fmoc-L-azetidine-2-carboxylic acid
OR962312-1G
APOSOR962312-1GEA
55.1
EUR
APOSOR962312-1G
(S)-N-Fmoc-azetidine-2-carboxylic acid 97%
(S)-N-Fmoc-azetidine-2-carboxylic acid
Formel:
C₁₉H₁₇NO₄ Molecular Weight: 323.348 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320843 CAS-Nummer: 136552-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...