N-Boc-3-cyano-L-phenylalanine 97%
Lieferant: Apollo Scientific
Synonyme:
Boc-Phe(3-CN)-OH
OR962365-5G
APOSOR962365-5GEA
74.1
EUR
APOSOR962365-5G
N-Boc-3-cyano-L-phenylalanine 97%
N-Boc-3-cyano-L-phenylalanine
Formel:
C₁₅H₁₈N₂O₄ Molecular Weight: 290.319 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00797560 CAS-Nummer: 131980-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...