2-((4-Cyanophenyl)amino)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: N-(4-CN-Phenyl)-Gly-OH, N-(4-Cyanophenyl)glycine

OR962439-500G
APOSOR962439-500GEA 104 EUR
APOSOR962439-500G
2-((4-Cyanophenyl)amino)acetic acid 95%
2-((4-Cyanophenyl)amino)acetic acid
Formel: C₉H₈N₂O₂
Molecular Weight: 176.1700 g/mol
Schmelzpunkt: 237 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01464022
CAS-Nummer: 42288-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR