N-Ethyl-2-nitroaniline 98%

Lieferant: Apollo Scientific

OR962442-5G
APOSOR962442-5GEA 171 EUR
APOSOR962442-5G
N-Ethyl-2-nitroaniline 98%
N-Ethyl-2-nitroaniline
Formel: C₈H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00007091
CAS-Nummer: 10112-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR