4-Hydroxy-6-nitroquinoline 96%

Lieferant: Apollo Scientific

OR962452-1G
APOSOR962452-1GEA 173 EUR
APOSOR962452-1G
4-Hydroxy-6-nitroquinoline 96%
4-Hydroxy-6-nitroquinoline
Formel: C₉H₆N₂O₃
Molecular Weight: 190,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024012
CAS-Nummer: 23432-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR