N-Butyl-4-nitroaniline 98%

Lieferant: Apollo Scientific

OR962453-5G
APOSOR962453-5GEA 62.5 EUR
APOSOR962453-5G
N-Butyl-4-nitroaniline 98%
N-Butyl-4-nitroaniline
Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024635
CAS-Nummer: 58259-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR