2-[(4-Nitrophenyl)amino]ethanol 95%

Lieferant: Apollo Scientific

OR962481-1G
APOSOR962481-1GEA 290 EUR
APOSOR962481-1G
2-[(4-Nitrophenyl)amino]ethanol 95%
2-[(4-Nitrophenyl)amino]ethanol
Formel: C₈H₁₀N₂O₃
Molecular Weight: 182,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156330
CAS-Nummer: 1965-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR