(4-Cyanophenoxy)acetic acid 96%

Lieferant: Apollo Scientific

Synonyme: (4-Cyanophenoxy)acetic Acid

OR962557-5G
APOSOR962557-5GEA 162 EUR
APOSOR962557-5G
(4-Cyanophenoxy)acetic acid 96%
(4-Cyanophenoxy)acetic acid
Formel: C₉H₇NO₃
Molecular Weight: 177.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01250758
CAS-Nummer: 1878-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR