5-Methylthiophene-2-carboxamide 95%

Lieferant: Apollo Scientific

OR962558-1G
APOSOR962558-1GEA 364 EUR
APOSOR962558-1G
5-Methylthiophene-2-carboxamide 95%
5-Methylthiophene-2-carboxamide
Formel: C₆H₇NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD01434337
CAS-Nummer: 57280-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR