1-(2-nitrophenyl)pyrrolidine 98%

Lieferant: Apollo Scientific

OR962562-100G OR962562-25G OR962562-5G
APOSOR962562-100GEA 632 EUR
APOSOR962562-100G APOSOR962562-25G APOSOR962562-5G
1-(2-nitrophenyl)pyrrolidine 98%
Formel: C₁₀H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01604393
CAS-Nummer: 40832-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR