N-Boc-4-carboxymethoxypiperidine 97%

Lieferant: Apollo Scientific

Synonyme: N-Boc-4-Carboxymethoxypiperidine, (1-Boc-4-piperidinyloxy)acetic acid

OR962583-1G
APOSOR962583-1GEA 120 EUR
APOSOR962583-1G
N-Boc-4-carboxymethoxypiperidine 97%
N-Boc-4-carboxymethoxypiperidine
Formel: C₁₂H₂₁NO₅
Molecular Weight: 259.30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02178987
CAS-Nummer: 161948-70-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR