2-(Cyclohexyloxy)acetic acid 98%
Lieferant: Apollo Scientific
OR962599-5G
APOSOR962599-5GEA
68.3
EUR
APOSOR962599-5G
2-(Cyclohexyloxy)acetic acid 98%
2-(Cyclohexyloxy)acetic acid
Formel:
C₈H₁₄O₃ Molecular Weight: 158.195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001653 CAS-Nummer: 71995-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...